Key Documents
MT51340336
Mettler-Toledo Economic & Easy conductivity sensor
with NTC30 temp. probe, 2 stainless steel poles, PVC and stainless steel shaft
About This Item
UNSPSC Code:
39121436
NACRES:
NB.61
material
PVC and stainless steel shaft
description
Mini-DIN connectior
conductivity - 0.01 μS/cm - 500 μS/cm
feature
2 stainless steel poles
manufacturer/tradename
Mettler-Toledo 51340336
General description
Low to medium conductivities
Robust conductivity probe.
Choose from one of the most recent versions:
Certificates of Analysis (COA)