Key Documents
MT51340335
Mettler-Toledo Economic & Easy conductivity sensor
with NTC30 temp. probe, 4 graphite poles, polycarbonate shaft (+ABS)
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
Pricing and availability is not currently available.
material
polycarbonate shaft (+ABS)
description
Mini-DIN connectior
conductivity - 0.01 μS/cm - 200 μS/cm
feature
4 graphite poles
manufacturer/tradename
Mettler-Toledo 51340335
temperature measuring range
0-80 °C
General description
Medium to high conductivities
Robust conductivity probe.
Choose from one of the most recent versions:
Certificates of Analysis (COA)