Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT51302582

Mettler-Toledo SevenGo Pro portable pH/Ion meter

SG8 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

description

Sensor inputs BNC (>1012 Ù); Cinch NTC 30 K? (both IP67)
Temperature range °C ATC: -5.0 ... 105.0; MTC: -30 ... 130 (+/- 0.2)

manufacturer/tradename

Mettler-Toledo 51302582

mV measuring range

-1999.9-1999.9, accuracy: ±0.1

battery power

4 × 1.5 V AA (or NiMH accumulators 1.3 V)

pH range

-2-19.999

Looking for similar products? Visit Product Comparison Guide

General description

Mettler Toledo SevenGo pro – Advanced pH/Ion meter for particularly demanding tasks coupled with exceptional ease of use. SevenGo pro pH/Ion features a lot of amazing operating functions, a large data memory for detailed GLP protocols, an infrared interface for data transfer to a printer or PC, bright backlighting, as well as a multitude of additional parameter-specific functions. SevenGo pro meters are truly indispensable for every analytical specialist, and in regulated working environments in the pharmaceutical, chemical, cosmetic and food manufacturing industries.

Features and Benefits - pH/Ion meter (IP67) for highest demands

  • Simple ion concentration determination
  • Time interval measurement with automatic data logging
  • Data memory for 200 GLP data points
  • Segmented or linear calibration as desired
  • 5-point calibration with choice of 6 predefined and one user-defined buffer set
includes meter, InLab® 413 SG IP67 rated "3-In-1" electrode (1.8 m fixed cable) complete with batteries, wrist strap, SevenGo clip, buffer sachets (2 each ; 4.01, 7.00 and 9.21), operating instructions, test certificate and declaration of conformity.

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.
SevenGo is a trademark of Mettler-Toledo, Inc.
SevenGo Pro is a trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service