Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT51302401

Mettler Toledo InLab® 717 conductivity probe

designed for use in connection with a titrator only

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115609
NACRES:
NB.79
Pricing and availability is not currently available.

material

glass shaft

description

4-pole platinum cell
mini DIN

manufacturer/tradename

Mettler-Toledo 51302401

conductivity measuring range

10-500,000 μS

L × W

120 mm × 12 mm

General description

Conductivity probe for titrations in aqueous or non-aqueous media in the conductivity range from 10 μS to 500 mS.

  • Glass 4 pole cell
  • Integrated connection cable, 1.0 m.
  • Integrated NTC temperature probe

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service