Key Documents
MT30034473
Mettler Toledo SevenExcellence conductivity expansion unit
About This Item
UNSPSC Code:
41115603
NACRES:
NB.61
Pricing and availability is not currently available.
manufacturer/tradename
Mettler-Toledo 30034473
General description
Can be used to expand any SevenExcellence meter to measure conductivity.
SevenExcellence fits three modules in any composition and order.
It offers the highest possible
flexibility as you can expand the instrument at any time, adding new measurement parameters.
Legal Information
SevenExcellence is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)