Key Documents
MT30019033
Mettler-Toledo SevenCompact conductivity meters S230
conductivity S230 kit, meter only, AC/DC input 230 V AC, universal plug set
About This Item
UNSPSC Code:
41115609
Pricing and availability is not currently available.
description
Calibration 13 pre-defined and 1 user-defined standard;
Conductivity range 0.001 ?S/cm to 1000 mS/cm, autoscaling (+/-0.5%)
Resistivity range 0.00 ...100 MOhm*cm, autoscaling
manufacturer/tradename
Mettler-Toledo 30019033
TDS measuring range
0-1000 g/L
salinity measuring range
0-80 (psu)
General description
Includes meter and electrode holder only.
Mettler Toledo SevenCompact conductivity meter S230 is the perfect choice for a wide range of applications without overstretching your budget. Not only does it measure conductivity but also various other parameters such as salinity, resistivity, total dissolved solids and conductivity ash.
The included electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample. This allows for faster measurements and reduces the risk of the sample vessel tipping over and/or damage the sensor.
SevenCompact gives you the choice to view all measurement information on the display or to focus on the essentials with the uFocus view. Switching between the different display layouts is easy, requiring only a single keypress.
The new color display with its well-designed icons and menu settings in 10 languages makes operation genuinely intuitive. Applications range from routine measurements through sample analysis, data handling and data archiving in compliance with GLP regulations. Simply innovative design that meets the requirements for a universal, easy-to-operate conductivity meter.
  • User friendly instrument for demanding users
  • Security and high reproducibility thanks to Intelligent Sensor Management (ISM®)
  • Improved quality of measurements through professional calibration support
  • Comprehensive service package including IQ/OQ
  • Integrated USB and RS232 interfaces for data exchange
Features and Benefits
Prominent color display
The 4.3 inch high-resolution color display with its large digits and well arranged icons makes relevant information identifiable at a glance.
Two display layouts
Switch with a single keypress between the full-information screen or the uFocus layout, in which you will not be distracted by information of less importance.
IntelligentSensor Management (ISM)
ISM helps eliminate mistakes by automatic identification of the sensor upon connection and ensures that the most recent calibration is used for future measurements.
Flexible calibration
For calibration you can choose between 13 pre-defined standards, set-up a user-defined standard or enter the cell constant manually in case you know its exact value.
Connectivity and peripherals
USB and RS232 interfaces allow you to connect various peripherals from METTLER TOLEDO simultaneously, increasing the possibilities of your instrument.
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
SevenCompact is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)