Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT30019029

Mettler Toledo SevenCompact pH/Ionmeters S220

pH/ion meter S220 kit, AC/DC input 230 V AC, universal plug set

Synonym(s):

ph meter, sevencompact ph meter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.61
Pricing and availability is not currently available.

description

Concentration range 1.00E-9 to 9.99E+9 (+/- 0.5%)

manufacturer/tradename

Mettler-Toledo 30019029

mV measuring range

-2000-2000, accuracy: ±0.2

temperature measuring range

-30-130 °C, accuracy: ±0.1

pH range

-2-20

Looking for similar products? Visit Product Comparison Guide

General description

Includes instrument, electrode holder, protective cover, operating instructions, quick guide, declaration of conformity and test certificate, InLab® Expert Pro-ISM, guide to pH measurements and 2 buffer sachets for pH 4.01, 7.00, 9.21 and 10.00.
Mettler Toledo SevenCompact pH/Ion meter S220 is the perfect choice for a wide range of applications without overstretching your budget. It is much more than a pH meter: it can also measure ORP and ion concentration in various units.

The included electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample. This allows for faster measurements and reduces the risk of the sample vessel tipping over and/or damage the sensor.

SevenCompact gives you the choice to view all measurement information on the display or to focus on the essentials with the uFocus view. Switching between the different display layouts is easy, requiring only a single keypress.

The new color display with its well-designed icons and menu settings in 10 languages makes operation genuinely intuitive. Applications range from routine measurements through sample analysis, data handling and data archiving in compliance with GLP regulations. Simply innovative design that meets the requirements for a universal, easy-to-operate pH/Ion meter.

  • User friendly instrument for demanding users
  • Security and high reproducibility thanks to Intelligent Sensor Management (ISM®)
  • Improved quality of measurements through professional calibration support
  • Comprehensive service package including IQ/OQ
  • Integrated USB and RS232 interfaces for data exchange


Features and Benefits
Prominent color display
The 4.3 inch high-resolution color display with its large digits and well arranged icons makes relevant information identifiable at a glance.

Two display layouts
Switch with a single keypress between the full-information screen or the uFocus layout, in which you will not be distracted by information of less importance.

IntelligentSensor Management (ISM)
ISM helps eliminate mistakes by automatic identification of the sensor upon connection and ensures that the most recent calibration is used for future measurements.

Automatic buffer recognition system
The automatic buffer recognition system allows you to choose the order in which to use buffers when calibrating. This way you quickly get through your routine calibrations without unnecessary error messages.

Connectivity and peripherals
USB and RS232 interfaces allow you to connect various peripherals from METTLER TOLEDO simultaneously, increasing the possibilities of your instrument.

Legal Information

ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
SevenCompact is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service