Key Documents
MT30014160
Mettler Toledo InLab® 725 conductivity probe
Designed for bio-ethanol analysis
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
Pricing and availability is not currently available.
material
glass shaft
description
2-pole platinum cell
cell constant 0.1 cm-1
manufacturer/tradename
Mettler-Toledo 30014160
L × W
120 mm × 12 mm
cable L
1 m
General description
A platinum 2-plate conductivity probe with integrated temperature sensor and fixed cable.
Designed for use with METTLER TOLEDO bench meters SevenExcellence and SevenCompact for determination of conductivity in bio-ethanol.
These two meters supports special units required for bio-ethanol analysis. This means unit inter-conversion and recalculation is no longer necessary.
The integrated temperature sensor avoids the need for a separate temperature probe when compensating for the difference in temperature between the sample and reference temperature.
Two-plate design provides a steady signal for accurate measurement of low conductivity samples.
  • Glass body is resistant to corrosive and organic sample types.
  • Two plate design
  • Integrated temperature sensor
  • Glass body
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
SevenCompact is a trademark of Mettler-Toledo, Inc.
SevenExcellence is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)