Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MIDA017

Sigma-Aldrich

5-Bromo-4-methoxy-2-TBDPS(benzyloxy)-boronic acid MIDA ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$36.70

$36.70


Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
$36.70

About This Item

Empirical Formula (Hill Notation):
C29H33BBrNO6Si
Molecular Weight:
610.38
UNSPSC Code:
12352108
PubChem Substance ID:

$36.70


Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

form

solid

SMILES string

COc1cc(OC[Si](c2ccccc2)(c3ccccc3)C(C)(C)C)c(cc1Br)B4OC(=O)CN(C)CC(=O)O4

InChI

1S/C29H33BBrNO6Si/c1-29(2,3)39(21-12-8-6-9-13-21,22-14-10-7-11-15-22)20-36-25-17-26(35-5)24(31)16-23(25)30-37-27(33)18-32(4)19-28(34)38-30/h6-17H,18-20H2,1-5H3

InChI key

PJKLECQBURGBHI-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service