Key Documents
MDFPFNB-M
Mediflex Gloves, blue nitrile, powder free
size M
About This Item
UNSPSC Code:
46181504
material
blue nitrile
size
M
description
textured
sterility
non-sterile
feature
latex free
powder-free
packaging
box of 100 gloves
L
240 mm
General description
Powder free blue nitrile glove. Non sterile, ambidextrous, latex
free, 240mm cuff, textured. Single use only.
Choose from one of the most recent versions:
Certificates of Analysis (COA)