Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8606

Sigma-Aldrich

Methanesulfonic acid

99%

Synonym(s):

Methanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3H
CAS Number:
Molecular Weight:
96.11
Beilstein/REAXYS Number:
1446024
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

3.3 (vs air)

Quality Level

vapor pressure

1 mmHg ( 20 °C)

assay

99%

form

liquid

impurities

<2 ppm methyl ester (a cancer suspect agent!)

refractive index

n20/D 1.429 (lit.)

bp

167 °C/10 mmHg (lit.)

mp

17-19 °C (lit.)

density

1.481 g/mL at 25 °C (lit.)

SMILES string

O=S(O)(C)=O

InChI

1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)

InChI key

AFVFQIVMOAPDHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

372.2 °F - closed cup

flash_point_c

189 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service