Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M47809

Sigma-Aldrich

3-Methylglutaric anhydride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
Beilstein/REAXYS Number:
114457
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

98%

bp

180-182 °C/25 mmHg (lit.)

mp

42-46 °C (lit.)

SMILES string

CC1CC(=O)OC(=O)C1

InChI

1S/C6H8O3/c1-4-2-5(7)9-6(8)3-4/h4H,2-3H2,1H3

InChI key

MGICRVTUCPFQQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maiko Miyazaki et al.
Bioconjugate chemistry, 29(1), 44-55 (2017-12-01)
For the enhancement of therapeutic effects and reduction of side effects derived from anticancer drugs in cancer chemotherapy, it is imperative to develop drug delivery systems with cancer-specificity and controlled release function inside cancer cells. pH-sensitive liposomes are useful as
Atsushi Harada et al.
Journal of biomaterials science. Polymer edition, 28(10-12), 1025-1035 (2017-04-27)
The properties of stimuli-responsive polymers change significantly with changes to their environment, such as temperature and pH. This behavior can be utilized for the preparation of stimuli-responsive carriers for efficient cytosolic delivery of active drugs. Among the possible environmental conditions
Peter A Beckmann et al.
Solid state nuclear magnetic resonance, 85-86, 1-11 (2017-03-07)
We report a variety of experiments and calculations and their interpretations regarding methyl group (CH
Eiji Yuba et al.
Pharmaceutics, 12(8) (2020-08-17)
Specific delivery to antigen presenting cells (APC) and precise control of the intracellular fate of antigens are crucial to induce cellular immunity that directly and specifically attacks cancer cells. We previously achieved cytoplasmic delivery of antigen and activation of APC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service