Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M46594

Sigma-Aldrich

1-Methylfluorene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12
CAS Number:
Molecular Weight:
180.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

84-86 °C (lit.)

SMILES string

Cc1cccc-2c1Cc3ccccc-23

InChI

1S/C14H12/c1-10-5-4-8-13-12-7-3-2-6-11(12)9-14(10)13/h2-8H,9H2,1H3

InChI key

GKEUODMJRFDLJY-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilberto Vasconcelos Villar et al.
Bulletin of environmental contamination and toxicology, 101(3), 349-357 (2018-08-08)
Polydimethylsiloxane (PDMS) is a polymer material with high absorptive properties increasingly used as a passive environmental sampler for persistent organic compounds. However, the partitioning behavior of hydrophobic chemicals to PDMS remains largely unknown. Organochlorines (OCs) and polycyclic aromatic hydrocarbons (PAHs)
J E Rice et al.
Journal of toxicology and environmental health, 21(4), 525-532 (1987-01-01)
The genotoxicity of 1-methylpyrene, 1,6-dimethylpyrene, 1-methylfluorene, 9-methylfluorene, and 1,9-dimethylfluorene was evaluated in the hepatocyte primary culture/DNA repair test, and the tumorigenic potency of these compounds was tested by bioassay in newborn mice. With the exception of 1-methylfluorene, all of these
Ifeoluwa Idowu et al.
Rapid communications in mass spectrometry : RCM, 32(3), 277-287 (2017-11-28)
There is a need for a validated method to improve detection limits and simultaneously quantify polycyclic aromatic compounds (PACs, both parent and alkylated homologues) in biota by gas chromatography/tandem mass spectrometry because of their environmental significance. The validation of the
Jaime Rodríguez-Estival et al.
Environmental toxicology and chemistry, 34(12), 2884-2897 (2015-07-04)
Increasing activity of oil sands extraction and processing in northern Alberta is marked by ongoing controversy about the nature and extent of associated environmental impacts. Bitumen contains a mixture of toxic chemicals, including metals and residual polycyclic aromatic hydrocarbons (PAHs)
Gail E Bragin et al.
Environmental toxicology and chemistry, 35(12), 2948-2957 (2016-05-07)
Because of the large number of possible aromatic hydrocarbon structures, predictive toxicity models are needed to support substance hazard and risk assessments. Calibration and evaluation of such models requires toxicity data with well-defined exposures. The present study has applied a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service