Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M31600

Sigma-Aldrich

3-Methylbenzyl cyanide

98%

Synonym(s):

3-Methylphenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2CN
CAS Number:
Molecular Weight:
131.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.52 (lit.)

bp

240-241 °C (lit.)

density

1.002 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(CC#N)c1

InChI

1S/C9H9N/c1-8-3-2-4-9(7-8)5-6-10/h2-4,7H,5H2,1H3

InChI key

WOJADIOTNFDWNQ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service