Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2501

Sigma-Aldrich

5-Methoxyindole-2-carboxylic acid ethyl ester

crystalline

Synonym(s):

2-Carbethoxy-5-methoxyindole, 2-Ethoxycarbonyl-5-methoxyindole, Ethyl 5-methoxyindole-2-carboxylate, NSC 30929

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO3
CAS Number:
Molecular Weight:
219.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

crystalline

color

yellow to orange

storage temp.

−20°C

SMILES string

CCOC(=O)c1cc2cc(OC)ccc2[nH]1

InChI

1S/C12H13NO3/c1-3-16-12(14)11-7-8-6-9(15-2)4-5-10(8)13-11/h4-7,13H,3H2,1-2H3

InChI key

NPIUAXNFAUGNHP-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Ligands with binding specificity towards the GABA receptor
  • Sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes
  • Anticancer agents
  • Integrase strand-transfer inhibitors (INSTIs)
  • Inhibitor of Proliferation of Colon Cancer Cells
  • Isomeridianin G as GSK-3? inhibitors
  • HIV-1 integrase inhibitors
  • Inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service