Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LR60000-HH

Sigma-Aldrich

Oxidizer 0.02M

Synonym(s):

Oxidizer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

type

for DNA synthesis

product line

Proligo Reagents

form

liquid

packaging

pkg of 4 × 2.5 L (LR60250-04)

concentration

3.9-4.7 g/L iodine (titration)

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acetonitrile / Water / Pyridine / Iodine 90.54 / 9.05 / 0.41 / 0.43 (v/v/v/W)

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service