Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L880045-HH

DCI Activator

0.25M, configured for PerkinElmer, configured for ABI, configured for Polygen

Synonym(s):

DCI activator solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 6 × 450 mL (L880045-06)

concentration

29-32 g/L (Dicyanoimidazole, titration)

impurities

≤30 ppm water content (Karl Fischer)

color

clear

compatibility

configured for ABI
configured for PerkinElmer
configured for Polygen

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Dicyanoimidazole / Acetonitrile 3/100 (w/v)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service