Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L754021

Sigma-Aldrich

PSiF-DBT

AldrichCPR, Mw 10,000-80,000

Synonym(s):

Poly[2,7-(9,9-dioctyl-dibenzosilole)-alt-4,7-bis(thiophen-2-yl)benzo-2,1,3-thiadiazole], Poly[2,1,3-benzothiadiazole-4,7-diyl-2,5-thiophenediyl(9,9-dioctyl-9H-9-silafluorene-2,7-diyl)-2,5-thiophenediyl]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C42H46N2S3Si)n
CAS Number:
UNSPSC Code:
12352200

mol wt

Mw 10,000-80,000

loss

0.5 wt. %, 300 °C

λmax

655 nm in methylene chloride
703 nm in methylene chloride

orbital energy

HOMO 5.39 eV (CV)

OPV device performance

ITO/PEDOT:PSS/PSiF-DBT:PC61BM (1:2)/Al

  • Short-circuit current density (Jsc): 9.5 mA/cm2
  • Open-circuit voltage (Voc): 0.9 V
  • Fill Factor (FF): 0.51
  • Power Conversion Efficiency (PCE): 5.4 %

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance polymer heterojunction solar cells of a polysilafluorene derivative
Wang, E.; Wang, L.; Lan, L.; et al.
Applied Physics Letters, 92, 033307-033307 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service