Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L512087

Sigma-Aldrich

PPT

AldrichCPR

Synonym(s):

2,8-Bis(diphenylphosphoryl)dibenzo[b,d]thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H26O2P2S
CAS Number:
Molecular Weight:
584.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O=P(C1=CC=CC=C1)(C2=CC=CC=C2)C(C=C3)=CC4=C3SC5=C4C=C(P(C6=CC=CC=C6)(C7=CC=CC=C7)=O)C=C5

InChI

1S/C36H26O2P2S/c37-39(27-13-5-1-6-14-27,28-15-7-2-8-16-28)31-21-23-35-33(25-31)34-26-32(22-24-36(34)41-35)40(38,29-17-9-3-10-18-29)30-19-11-4-12-20-30/h1-26H

InChI key

ZCJJIQHVZCFSGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takekazu Iwata et al.
The Annals of thoracic surgery, 102(6), 1905-1910 (2016-08-23)
Acute exacerbation of idiopathic pulmonary fibrosis (AE-IPF) is a life-threatening complication of lung cancer operation for patients with IPF, and no effective prophylaxis has ever been reported. In this study, we investigate the effect of perioperative treatment with an anti-IPF
Ester C Sabino et al.
The Journal of infectious diseases, 213(5), 694-702 (2016-02-26)
A linked donor-recipient study was conducted during epidemics in 2 cities in Brazil to investigate transfusion-transmitted (TT) dengue virus (DENV) by DENV RNA-positive donations. During February-June 2012, samples were collected from donors and recipients and retrospectively tested for DENV RNA
Sergi Soriano et al.
Scientific reports, 9(1), 10306-10306 (2019-07-18)
17β-Estradiol mediates the sensitivity to pain and is involved in sex differences in nociception. The widespread environmental disrupting chemical bisphenol A (BPA) has estrogenic activity, but its implications in pain are mostly unknown. Here we show that treatment of male
Joan O'Riordan et al.
Transfusion, 56(11), 2868-2876 (2016-08-16)
Hepatitis E virus (HEV) Genotype 3 (G3) infection is a zoonosis that may be transmitted during the acute phase by transfusion. The aim of this study was to determine the incidence of HEV and seroprevalence among Irish blood donors. Anonymized
Vladimir Vuksan et al.
European journal of nutrition, 58(3), 1237-1245 (2018-02-27)
Despite the lack of evidence, a growing number of people are using herbal medicine to attenuate the burden of diabetes. There is an urgent need to investigate the clinical potential of herbs. Preliminary observations suggest that American ginseng (Panax quinquefolius

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service