Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L511641

Sigma-Aldrich

5-Bromo-4-(trimethylsilyl)pyridin-3-yl trifluoromethanesulfonate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BrF3NO3SSi
Molecular Weight:
378.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

BrC1=CN=CC(OS(=O)(C(F)(F)F)=O)=C1[Si](C)(C)C

InChI

1S/C9H11BrF3NO3SSi/c1-19(2,3)8-6(10)4-14-5-7(8)17-18(15,16)9(11,12)13/h4-5H,1-3H3

InChI key

DFTOVRWLXREECB-UHFFFAOYSA-N

Packaging

Each AldrichCPR product will be packaged in one consolidated unit unless you specify multiple units at the time of order.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul H-Y Cheong et al.
Journal of the American Chemical Society, 132(4), 1267-1269 (2010-01-12)
Density functional theory computations reproduce the surprisingly high regioselectivities in nucleophilic additions and cycloadditions to 4,5-indolynes and the low regioselectivities in the reactions of 5,6-indolynes. Transition-state distortion energies control the regioselectivities, activating the 5 and 6 positions over the 4
Adam E Goetz et al.
Angewandte Chemie (International ed. in English), 51(11), 2758-2762 (2012-02-07)
Think before you act: a computational approach is reported for evaluating the synthetic potential of heterocyclic arynes. Routine and rapid calculations of arene dehydrogenation energies and aryne angle distortion predict the likelihood that a given hetaryne can be generated, as
Adam E Goetz et al.
Nature chemistry, 5(1), 54-60 (2012-12-19)
The pyridine heterocycle continues to play a vital role in the development of human medicines. More than 100 currently marketed drugs contain this privileged unit, which remains highly sought after synthetically. We report an efficient means to access di- and
G-Yoon J Im et al.
Journal of the American Chemical Society, 132(50), 17933-17944 (2010-12-01)
Efficient syntheses of 4,5-, 5,6-, and 6,7-indolyne precursors beginning from commercially available hydroxyindole derivatives are reported. The synthetic routes are versatile and allow access to indolyne precursors that remain unsubstituted on the pyrrole ring. Indolynes can be generated under mild
Sarah M Bronner et al.
Journal of the American Chemical Society, 133(11), 3832-3835 (2011-03-01)
We report the design and synthesis of an indolyne that displays a reversal in regioselectivity, in both nucleophilic addition and cycloaddition reactions, compared to typical 4,5-indolynes. Our approach utilizes simple computations to predict regioselectivity in reactions of unsymmetrical arynes. With

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service