Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L511528

Sigma-Aldrich

(S)-4-(tert-Butyl)-2-[4-(trifluoromethyl)pyridin-2-yl]-4,5-dihydrooxazole

Synonym(s):

(S)-5-CF3-pyrox-tBu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15F3N2O
Molecular Weight:
272.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

CC(C)(C)[C@H]1COC(=N1)c2ccc(cn2)C(F)(F)F

InChI

1S/C13H15F3N2O/c1-12(2,3)10-7-19-11(18-10)9-5-4-8(6-17-9)13(14,15)16/h4-6,10H,7H2,1-3H3/t10-/m1/s1

Inchi Key

AATATIAROAAKIN-SNVBAGLBSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service