Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L510378

Sigma-Aldrich

2,2′-Bis(trifluoromethyl)benzidine

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10F6N2
CAS Number:
Molecular Weight:
320.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

Nc1ccc(-c2ccc(N)cc2C(F)(F)F)c(c1)C(F)(F)F

InChI

1S/C14H10F6N2/c15-13(16,17)11-5-7(21)1-3-9(11)10-4-2-8(22)6-12(10)14(18,19)20/h1-6H,21-22H2

InChI key

NVKGJHAQGWCWDI-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sun Dal Kim et al.
Science advances, 4(10), eaau1956-eaau1956 (2018-11-07)
The key component currently missing for the next generation of transparent and flexible displays is a high-performance polymer material that is flexible, while showing optical and thermal properties of glass. It must be transparent to visible light and show a
Yingyi Ma et al.
Chemistry, an Asian journal (2018-06-28)
A honeycomb-patterned porous film was conveniently prepared by means of the microemulsion method and then used as an effective coating for improving the dielectric properties and water resistance of polyimide (PI) film while maintaining its mechanical properties. The dielectric properties
Mei Zhang et al.
Polymers, 12(7) (2020-07-16)
Transparent polyimides (PI) films with outstanding overall performance are attractive for next generation optoelectronic and microelectronic applications. Semi-alicyclic PIs derived from alicyclic dianhydrides and aromatic diamines have proved effective to prepare transparent PIs with high transmittance. To optimize the combined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service