All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H5NO5
CAS Number:
Molecular Weight:
183.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
SMILES string
Oc1cc(nc(c1)C(O)=O)C(O)=O
InChI
1S/C7H5NO5/c9-3-1-4(6(10)11)8-5(2-3)7(12)13/h1-2H,(H,8,9)(H,10,11)(H,12,13)
InChI key
XTLJJHGQACAZMS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[Antiviral activity of chelidamic acid derivatives].
V E Iavorovskaia et al.
Voprosy virusologii, 29(3), 361-363 (1984-05-01)
Ahmed Moussa et al.
Chirality, 21(5), 497-506 (2008-08-14)
The perturbation of the racemic equilibrium of luminescent D3 terbium(III) complexes with chelidamic acid (CDA), a hydroxylated derivative of 2,6-pyridine-dicarboxylic acid (DPA), by added chiral biomolecules such as L-amino acids has been studied using circularly polarized luminescence and 13C NMR
[Use of the radial hemolysis reaction for titrating antirabies sera].
G N Zgurskaia et al.
Voprosy virusologii, 29(3), 360-361 (1984-05-01)
M Searcey et al.
Anti-cancer drug design, 13(8), 837-855 (1999-05-21)
We have explored the potential antitumour activity of DNA-intercalating free radical generators based on compounds constructed from 9-anilinoacridine and chelidamic acid as an iron (II) binding centre. Here we describe their synthesis, DNA cleaving ability and activity against a panel
Elvir Ramić et al.
The journal of physical chemistry. B, 110(41), 20655-20663 (2006-10-13)
Multifrequency electron paramagnetic resonance (EPR) and electron nuclear double resonance (ENDOR) techniques were used to obtain structural information about the copper(II)-chelidamate complex. Well-resolved nitrogen ENDOR spectra could be recorded from solid solution samples by using selective excitation of spin packets.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service