Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L380018

Sigma-Aldrich

DCI Activator

0.25M, configured for ABI

Synonym(s):

DCI activator solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 6 × 180 mL L380018-06

concentration

29-32 g/L (titration)

impurities

≤30 ppm water content (Karl Fischer)

color

clear

compatibility

configured for ABI

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

41.0 °F

flash_point_c

5 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service