Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L260000-HH

Sigma-Aldrich

Oxidizer 0.02M

configured for ABI 3900 synthesizer

Synonym(s):

Oxidizer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 6 × 450 mL L260045-06

concentration

5.4-6.6 g/L iodine (titration)

compatibility

configured for ABI 3900 synthesizer

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Tetrahyrofurane/Water/Pyridine/Iodine 66/12/22/0.6 (v/v/v/w)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

50.0 °F

flash_point_c

10 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service