Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L093000

Sigma-Aldrich

Pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein/REAXYS Number:
103233
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
bp:
115 °C (lit.)
vapor pressure:
10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

type

for DNA synthesis

product line

Proligo Reagents

form

liquid

autoignition temp.

899 °F

expl. lim.

12.4 %

storage condition

dry at room temperature

impurities

≤100 ppm water content (Karl Fischer)
conforms Absence of Amine (titration)

color

clear

refractive index

n20/D 1.509 (lit.)

pH

8.5 (25 °C, 15.82 g/L)

bp

115 °C (lit.)

mp

−42 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

68.0 °F - closed cup

flash_point_c

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Christopher T Krüger et al.
Archives of toxicology, 89(12), 2429-2443 (2014-11-25)
The X-linked PIG-A gene is involved in the biosynthesis of the cell surface anchor GPI, and its inactivation may serve as a new marker for mutagenicity. The in vivo PIG-A gene mutation assay is currently being validated by several groups.
Danilo M Daloso et al.
Plant, cell & environment, 38(11), 2353-2371 (2015-04-15)
Transcriptomic and proteomic studies have improved our knowledge of guard cell function; however, metabolic changes in guard cells remain relatively poorly understood. Here we analysed metabolic changes in guard cell-enriched epidermal fragments from tobacco during light-induced stomatal opening. Increases in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service