Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L040000-HH

Sigma-Aldrich

Cap A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 4 × 2 L (L040200-04)
pkg of 1 × 2.5 L (L040250-01)
pkg of 4 × 2.5 L (L040250-04)
pkg of 1 × 4 L (L040400-01)
pkg of 4 × 4 L (L040400-04)

concentration

88.7-108.4 g/L (Acetic Anhydride content, titration)

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acetic Anhydride / Tetrahydrofurane 9.1/90.9 (v/v)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

6.1 °F

flash_point_c

-14.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service