Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L020080

Sigma-Aldrich

TCA Deblock

configured for MerMade, configured for PerkinElmer, configured for Polygen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

type

for DNA synthesis
for DNA synthesis

product line

Proligo Reagents

storage condition

dry at room temperature

concentration

28-32 g/L (TCA content, titration)

impurities

≤200 ppm water content (Karl Fischer)

color

clear
clear

compatibility

configured for MerMade
configured for PerkinElmer
configured for Polygen

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Trichloroacetic acid / Dichloromethane 3/100 (v/v)

Other Notes

Deblock solution containing trichloroacetic acid (TCA) in dichloromethane removes the dimethoxytrityl (DMT) protecting group from the 5′hydroxyl moiety of nucleotides already incorporated into the growing nucleic acid, prior to the addition of the next phosphoramidite. Removal of the DMT allows the unprotected 5′hydroxyl moiety to react with a new phosphoramidite in a subsequent extension reaction.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service