Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L020000-HH

Sigma-Aldrich

TCA Deblock

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

packaging

pkg of 4 × 2.5 L (L020250-04)
pkg of 1 × 25 L (L022500-01)
pkg of 4 × 4 L (L020400-04)

concentration

28-32 g/L (TCA content, titration)

impurities

≤200 ppm water content (Karl Fischer)

color

clear

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Trichloroacetic acid / Dichloromethane 3/100 (v/v)

Other Notes

Deblock solution containing trichloroacetic acid (TCA) in dichloromethane removes the dimethoxytrityl (DMT) protecting group from the 5′hydroxyl moiety of nucleotides already incorporated into the growing nucleic acid, prior to the addition of the next phosphoramidite. Removal of the DMT allows the unprotected 5′hydroxyl moiety to react with a new phosphoramidite in a subsequent extension reaction.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

212.0 °F - closed cup

flash_point_c

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service