Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L0200002-HH

Sigma-Aldrich

TFA Deblock Toluene

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.21

product line

Proligo Reagents

form

liquid

packaging

pkg of 4 × 2.5 L

Looking for similar products? Visit Product Comparison Guide

Components

7.4 / 1000 Trifluoracetic acid / Toluene (v/v)

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F

flash_point_c

4 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service