Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

KITALYSIS-SCAV

Sigma-Aldrich

KitAlysis High-Throughput Metal Scavenger Screening Kit-Pack of 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

reaction suitability

core: palladium

storage temp.

2-8°C

Application

Screening kit for metal scavenger reactions. A Step-by-step user guides is provided in the below hyperlinks.
Step-by-Step Guide for KitAlysis High-Throughput Metal Scavenger Reaction Screening Kit.

Each kit contains 2 customizable screening sets that include the following components:
  • 24 (12 x 2) pre-weighed scavengers / resins in glass vials topped with cap mat.

Features and Benefits

  • Designed and tested by synthetic chemists.
  • Easily run 24-microscale reaction in parallel.
  • Useful for broad-spectrum metal removal, impurity removal, and color treatment.

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service