Key Documents
KF1842047
Whatman® glass microfiber filters Multigrade GMF 150
Synonym(s):
Whatman filter, glass microfiber filters
About This Item
UNSPSC Code:
41104918
Pricing and availability is not currently available.
material
borosilicate glass membrane
feature
hydrophilic
manufacturer/tradename
Whatman Article No. 28418508 (US reference)
pore size
1.2 μm
General description
The Whatman GMF 150 is a multilayer glass microfiber filter with a coarse top layer (10 μm) meshed with a finer layer of 1.0 μm or 2.0 μm. Manufactured from 100% borosilicate glass microfiber, the filter is binder free. It is the ideal prefilter for higher particulate loading capacity with faster flow rates.
The GMF 150 allows for:
  • Higher particulate loading capacity
  • Faster flow rate
  • Extended life of filter
  • Multiple porosities, greater filtration efficiency
The GMF 150 represents a new dimension in separation science leading to faster and more cost-effective filtration. In application, the GMF 150* traps larger particles in the pores or on the surface of the coarse layer while the medium sized particles are caught in the interface meshing. The smaller particles are netted in the interstices of the fine layer.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)