JRD0804
3-(4-(4-Bromophenoxy)phenyl)-1H-pyrazole-5-carbohydrazide
Select a Size
$639.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$639.00
Notify Me
Get notified when this item is ready to ship via email.
form
solid
SMILES string
BrC(C=C1)=CC=C1OC2=CC=C(C=C2)C3=NNC(C(NN)=O)=C3
InChI
1S/C16H13BrN4O2/c17-11-3-7-13(8-4-11)23-12-5-1-10(2-6-12)14-9-15(21-20-14)16(22)19-18/h1-9H,18H2,(H,19,22)(H,20,21)
InChI key
ZBKMZVHNNSIICB-UHFFFAOYSA-N
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service