Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA5024

Sigma-Aldrich

Acrylate-PEG7500-NHS

average Mn 7,500

Synonym(s):

Acrylate-PEG-NHS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2CHCOO(CH2CH2O)nCH2COOC4H4O2N
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 7,500

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions
reactivity: amine reactive

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Acrylate PEG NHS Ester. Light sensitive PEG, will crosslink with exposure to ultraviolet light.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dang, L.T.H., et al.
Biomaterials, 35(27) (2014)
Qi, H., et al.
Nature Communications, 4 (2013)
Hoffman, M.D., et al.
Acta Biomaterialia, 10(8) (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service