Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA4048

Sigma-Aldrich

Acrylate-PEG3500-Acrylate

average Mn 3,500

Synonym(s):

Acrylate-PEG-Acrylate, ACLT-PEG-ACLT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2CHCOO(CH2CH2O)nCH2CH2OOCCHCH2
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 3,500

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

General description

Acrylate-PEG-Acrylate. PEG crosslinker used in vinyl polymerization or co-polymerization.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service