Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA3004

Sigma-Aldrich

mPEG30K-Succinimidyl Carboxymethyl Ester

average Mn 30,000

Synonym(s):

mPEG-Succinimidyl Carboxymethyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O(CH2CH2O)nCH2COOC4H4O2N
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 30,000

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Methoxy PEG NHS Ester (SCM PEG). Amine reactive PEG towards the amino groups of lysine(s) on proteins or other biologics; reaction occurs at room temperature in <1hr at pH 7-8. Stable linker between PEG and NHS ester.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao, G., et al.
Langmuir, 30(23) (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service