All Photos(1)
About This Item
Linear Formula:
CH3O(CH2CH2O)5CH2CH2OCOCHCH2
UNSPSC Code:
12352200
NACRES:
NA.23
Recommended Products
form
solid
storage temp.
−20°C
Related Categories
Application
Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization
Legal Information
Product of JenKem Technology
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service