Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA12001

Sigma-Aldrich

HO-PEG4-Propionic acid

Synonym(s):

HO-PEG4-PA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)4CH2CH2COOH
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

reagent type: chemical modification reagent
reactivity: amine reactive

reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

SMILES string

OCCOCCC(O)=O

InChI

1S/C5H10O4/c6-2-4-9-3-1-5(7)8/h6H,1-4H2,(H,7,8)

InChI key

GTIFLSYPOXYYFG-UHFFFAOYSA-N

General description

Monodisperse PEG. COOH group is stable and can be activated.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service