JKA10041
8arm-PEG10K 7arm-OH, 1arm-COOH, tripentaerythritol core
average Mn 10,000
Synonym(s):
8arm-PEG 7arm-OH, 1arm-COOH, tripentaerythritol core, 8arm PEG, 7arm-Hydroxyl, 1arm-Carboxyl
About This Item
form
solid
mol wt
average Mn 10,000
reaction suitability
reactivity: amine reactive
polymer architecture
shape: 8-arm
functionality: heterofunctional
Storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service