Y-shaped Propion Aldehyde PEG. Reactive PEG for N-terminal amine in the presence of a reducing reagent; less selective but more reactive compared with Y-AALD-40K; reacts at pH 5-8. JenKem Technology′s patented products, Y-shape PEG derivatives, also known as branched PEG derivatives, contain two linear methoxy PEG chains attached to a central active core. JenKem proprietary Y-shape PEGs are more selective, due to their sterically bulky structure.
Application
Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization
Legal Information
Product of JenKem Technology
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.