Key Documents
HSWNH25112
HSW FINE-JECT hypodermic needles, disposable
gauge 25, L 1 1/2 in., orange, sterile
About This Item
UNSPSC Code:
42142615
material
orange
description
thin wall
sterility
sterile
manufacturer/tradename
Henke Sass Wolfe
L
1 1/2 in.
gauge
25
General description
  • Disposable, sterile, FDA registered
  • ISO 9626/1991 stainless steel
  • Color-coded polypropylene hub
  • Protective polypropylene cap
  • Siliconized needle tube
  • Pyrogen-free and non-toxic
  • 5-year shelf life
Choose from one of the most recent versions:
Certificates of Analysis (COA)