Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF97462806

Nickel-aluminum alloy, Ni79Al21

powder, 150micron max. particle size, alloy pre-cursor, 200g

Synonym(s):

NI226010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300

form

powder

manufacturer/tradename

Goodfellow 974-628-06

weight

200 g

max. part. size

150 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service