Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF97027443

Platinum

foil, 0.2m coil, thickness 0.025mm, coil width 1.2mm, as rolled, 99.95%

Synonym(s):

Platinum, PT000235

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$571.00

$571.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$571.00

About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

$571.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥99.95%

form

foil

manufacturer/tradename

Goodfellow 970-274-43

resistivity

10.6 μΩ-cm, 20°C

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalia Nayak et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 57(4), 483-489 (2002-10-04)
Heavy ion activation of natural gold foil with a 7Li beam results in the formation of carrier-free 199-201Pb radionuclides along with their corresponding decay products, 199-201Tl, in the matrix. At 49 MeV projectile energy, the production of 199Tl through its
Using platinum foil techniques to create natural tooth color.
Luke Kahng
Practical procedures & aesthetic dentistry : PPAD, 20(8), 479-481 (2008-11-11)
Martin Schuettler et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2009, 1612-1615 (2009-12-08)
An easy and fast method for fabrication of neural electrode arrays is the patterning of platinum foil and spin-on silicone rubber using a laser. However, the mechanical flexibility of such electrode arrays is limited by the integrated tracks that connect
Maurice Y Mommaerts et al.
Child's nervous system : ChNS : official journal of the International Society for Pediatric Neurosurgery, 19(4), 211-216 (2003-04-26)
Our hypothesis was that a neurocranial suture autograft will, when shielded from dura, grow and be incorporated into the calvarium. Growth was monitored by marker separation in three isohistogenic groups of rabbits, between postnatal days 9 and 90. In order
Minimally invasive makeover: platinum foil technique.
Steve Ballis et al.
Dentistry today, 29(2), 146-146 (2010-03-04)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service