No, this product is not a dead-end tube; it has both ends open.
GF96925393
Alumina
tube, outside diameter 6.0 mm, length 200 mm, wall thickness 1.0 mm
Synonym(s):
AL607120
About This Item
form
tubes
manufacturer/tradename
Goodfellow 969-253-93
L
200 mm
O.D.
6.0 mm
wall thickness
1.0 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hi, for the GF96925393 Al2O3 tube, outside diameter 6.0 mm, length 200 mm, wall thickness 1.0 mmm, is this tube with one dead-end? That's what I am looking for. Thanks!
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service