Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF95617020

Yttrium

wire reel, 0.05m, diameter 1.0mm, square section, 99.9%

Synonym(s):

Yttrium, Y 005111

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y
CAS Number:
Molecular Weight:
88.91
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

≥99.9%

form

wire

manufacturer/tradename

Goodfellow 956-170-20

resistivity

57 μΩ-cm, 20°C

bp

3338 °C (lit.)

mp

1522 °C (lit.)

density

4.469 g/mL at 25 °C (lit.)

SMILES string

[Y]

InChI

1S/Y

InChI key

VWQVUPCCIRVNHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiko Ishihara et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(8), 781-785 (2015-08-11)
With a view to enhance the sensitivity of analytical instruments used in the measurement of trace elements contained in a single cell, we have now equipped the previously reported micro-droplet injection system (M-DIS) with a desolvation system. This modified M-DIS
G Scholz et al.
Dalton transactions (Cambridge, England : 2003), 44(30), 13522-13529 (2015-07-03)
Nanoscopic yttrium acetate fluorides Y(CH(3)COO)(3-z)F(z) and yttrium oxide fluorides YO(3-z)/(2)F(z )were prepared with tunable Y/F molar ratios via the fluorolytic sol-gel route. All samples were characterized by X-ray diffraction, elemental analysis and thermal analysis. In addition, local structures of all
Ryan Hickey et al.
International journal of radiation oncology, biology, physics, 88(5), 1025-1031 (2014-03-26)
Radiosensitizing chemotherapy improves the outcomes in comparison with radiation alone for gastrointestinal cancers. The delivery of radiation therapy with yttrium90 ((90)Y) radioembolization, in combination with the radiosensitizing chemotherapeutic agent capecitabine, provides the opportunity to enhance the effects of radiation on
Dan Li et al.
Journal of nanoscience and nanotechnology, 14(6), 4207-4213 (2014-04-18)
YF3:Er3+ hollow nanofibers were successfully fabricated via fluorination of the relevant Y2O3:Er3+ hollow nanofibers which were obtained by calcining the electrospun PVP/[Y(NO3)3 + Er(NO3)3] composite nanofibers. The morphology and properties of the products were investigated in detail by X-ray diffraction
Hojjat Ahmadzadehfar et al.
Seminars in nuclear medicine, 40(2), 105-121 (2010-02-02)
Radioembolization (RE), also termed selective internal radiation therapy (SIRT), has been gradually introduced to the clinical arsenal of cytoreductive modalities in recent years. There is growing evidence for efficiency in liver tumors of various entities, with the most prominent ones

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service