Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF95347110

Nickel

tube, 500mm, outside diameter 0.762mm, inside diameter 0.204mm, wall thickness 0.279mm, hard, 99.5%

Synonym(s):

Nickel, NI007151

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

assay

≥99.5%

form

tubes

manufacturer/tradename

Goodfellow 953-471-10

resistivity

6.97 μΩ-cm, 20°C

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary A Molander et al.
Organic letters, 16(7), 1904-1907 (2014-03-29)
A general method for the alkenylation of alkyl electrophiles using nearly stoichiometric amounts of the air- and moisture-stable potassium organotrifluoroborates has been developed. Various functional groups were tolerated on both the nucleophilic and electrophilic partner. Reactions of highly substituted E-
Hao Yuan et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 72(4), 764-772 (2014-03-19)
In surgery-first accelerated orthognathic surgery, the clinical phenomenon of accelerated orthodontic tooth movement after osteotomy is a benefit compared with the conventional approach. However, because much of the literature on this phenomenon is based on empirical evidence and case reports
Som Niyogi et al.
Aquatic toxicology (Amsterdam, Netherlands), 150, 36-44 (2014-03-19)
The freshwater pulmonate snail, Lymnaea stagnalis, is the most sensitive aquatic organism tested to date for Ni. We undertook a series of experiments to investigate the underlying mechanism(s) for this observed hypersensitivity. Consistent with previous experiments, juvenile snail growth in
Gonensin O Bozdag et al.
Gene, 538(2), 251-257 (2014-02-04)
Nickel is an essential micronutrient due to its involvement in many enzymatic reactions as a cofactor. However, excess of this element is toxic to biological systems. Here, we constructed a cDNA library from Beta maritima and screened it in the
Egor Geist et al.
Natural product reports, 31(4), 441-448 (2014-02-28)
This Highlight covers the current status of relatively unexplored sp(3)-sp(3) cross-coupling reactions with particular focus on natural product and related syntheses.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service