Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF93059994

Copper-beryllium alloy, Cu98Be2

foil, 50mm disks, 0.05mm thickness, quarter hard

Synonym(s):

CU070301

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
31152304
NACRES:
NA.23

form

foil (disks - quarter hard)

manufacturer/tradename

Goodfellow 930-599-94

size × thickness

50 mm × 0.05 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service