GF92776809
Titanium-aluminum-vanadium alloy, Ti90Al6V4
sphere, 3mm diameter, 5.08μm sphericity, precision sphere, grade 200, 50pcs
Synonym(s):
TI016830
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352300
NACRES:
NA.23
form
spheres (grade 200)
quality
precision sphere
packaging
1 ea of 50 pieces
manufacturer/tradename
Goodfellow 927-768-09
diam.
3 mm
sphericity
5.08 μm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service