Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF87587959

Tantalum

wire reel, 1m, diameter 0.025mm, as drawn, 99.9%

Synonym(s):

Tantalum, TA005100, Ta

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$302.00

$302.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$302.00

About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

$302.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

vapor pressure

<0.01 mmHg ( 537.2 °C)

assay

≥99.9%

form

wire

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 875-879-59

resistivity

13.5 μΩ-cm, 20°C

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical use of porous tantalum in complex primary total knee arthroplasty.
Neil P Sheth et al.
American journal of orthopedics (Belle Mead, N.J.), 38(10), 526-530 (2009-12-17)
Brett Levine et al.
The Journal of the American Academy of Orthopaedic Surgeons, 14(12), 646-655 (2006-11-02)
Porous tantalum is an alternative metal for total joint arthroplasty components that offers several unique properties. Its high volumetric porosity (70% to 80%), low modulus of elasticity (3 MPa), and high frictional characteristics make it conducive to biologic fixation. Tantalum
[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
R M Rabinovich
Vestnik rentgenologii i radiologii, (1)(1), 77-79 (1981-01-01)
Xiao-kang Tang et al.
Zhongguo gu shang = China journal of orthopaedics and traumatology, 26(7), 617-620 (2013-10-19)
Incorrect treatment for femur head necrosis can cause collapse of femoral head and tresult in severe harm for the patients (especially for the patient with middle-aged and young). The structure and mechanics characteristics of tantalum rod is similar to bone
Ulrich Dorn et al.
The Journal of arthroplasty, 29(4), 831-835 (2013-10-09)
This study compared the corrosion behavior of tantalum-coated cobalt-chromium modular necks with that of titanium alloy modular necks at their junction to titanium-alloy femoral stem. Tests were performed in a dry assembly and two wet assemblies, one contaminated with calf

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service