Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF87362118

Tungsten

wire reel, 25m, diameter 0.015mm, gold plated, 99.95%

Synonym(s):

Tungsten, W 005220, W

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$440.00

$440.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$440.00

About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23

$440.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

assay

≥99.95%

form

wire

manufacturer/tradename

Goodfellow 873-621-18

resistivity

4.9 μΩ-cm, 20°C

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Boll et al.
Biological chemistry, 386(10), 999-1006 (2005-10-13)
The molybdenum enzymes 4-hydroxybenzoyl-CoA reductase and pyrogallol-phloroglucinol transhydroxylase and the tungsten enzyme acetylene hydratase catalyze reductive dehydroxylation reactions, i.e., transhydroxylation between phenolic residues and the addition of water to a triple bond. Such activities are unusual for this class of
Malcolm H Chisholm et al.
Chemical record (New York, N.Y.), 5(5), 308-320 (2005-10-08)
The reaction of M2(O2CBu(t))4 (M = Mo, W) with a dicarboxylic acid in toluene yields compounds of general formula [M2]-O2C-X-CO2-[M2] ([M2] = M2(O2CBu(t))3; X = conjugated spacer). The M2 units are electronically coupled via interactions between the M2 delta and
José L Domingo
Biological trace element research, 88(2), 97-112 (2002-09-26)
Tungstate is an oxyanion that has biological similarities to vanadate. In recent years, a number of studies have shown the antidiabetic effects of oral tungstate in animal models of diabetes. However, because of the tissue accumulation and potential toxicity derived
A Koutsospyros et al.
Journal of hazardous materials, 136(1), 1-19 (2005-12-14)
Since its discovery, tungsten, a transition element of Group VIb of the Periodic Table of Elements, and its compounds have been considered environmentally benign. Its presence in biological and drinking water samples in Fallon, Nevada, an acute lymphocytic leukemia cluster
Vernon G Thomas et al.
Ecotoxicology and environmental safety, 72(4), 1031-1037 (2009-02-24)
The toxicity of elemental tungsten released from discharged shot was assessed against previous studies that established a 1% toxic threshold for soil organisms. Extremely heavy theoretical shot loadings of 69,000shot/ha were used to generate estimated environmental concentrations (EEC) for two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service